Cerium molybdenum oxide ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13454-70-1
:
Formula:Ce₂(MoO₄)₃
MDL Number:MFCD00798520
:760.05 g/mol
Melting Pt:973 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together