Order Entry
Czech Republic
ContactUsLinkComponent
Tin oxide, antimony doped (∼30% Oxide) (w/w) polymeric precursor
Irritant
Tin oxide, antimony doped (∼30% Oxide) (w/w) polymeric precursor
Catalog # 39754.14
CAS Number:  
Irritant
Tin oxide, antimony doped (∼30% Oxide) (w/w) polymeric precursor
Catalog # 39754.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Density:1.61
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00011244