
Barium tetracyanoplatinate(II) tetrahydrate ≥99.99% (metals basis), Premion®
Catalog # 39463.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:508.542 g/mol
Merck Index:14,991
Melting Pt:100 °C (dec.)
Density:3.05
Storage Temperature:Ambient
EINECS:209-238-1
UN:1564
ADR:6.1,II
:GHS06
CAS Number:13755-32-3
Formula:BaPt(CN)₄.4H₂O
MDL Number:MFCD00014183