Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Přihlásit se
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dihydrogen hexafluorozirconate
Dihydrogen hexafluorozirconate 20% (w/w) in aqueous solution
Dihydrogen hexafluorozirconate 20% (w/w) in aqueous solution
39461.A1
:
Thermo Fisher Scientific
:
Dihydrogen hexafluorozirconate 20% (w/w) in aqueous solution
39461.A1
:
Thermo Fisher Scientific
:
Pk:
1 kg
Seq:
4024
:
:
1.52
:
234-666-0
:
8,II
:
205.215 Da
:
3264
:
GHS05
:
GHS07
:
Ambient
:
12021-95-3
:
H₂ZrF₆
:
MFCD00082965