Order Entry
Czech Republic
ContactUsLinkComponent
 
Nitrate, Ion chromatography standard solution, Specpure®, NO₃- 1000 µg/ml
 
Nitrate, Ion chromatography standard solution, Specpure®, NO₃- 1000 µg/ml
 

 

  • MDL No.:
    MFCD00144920
  • Physical form:
    Liquid
  • Molecular formula:
    NaNO3 in H2O

 

Frequently Bought Together