
Sodium hexachloroosmate(IV) dihydrate Os ≥38,7%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1307-81-9
:
Formula:Cl₆Na₂Os·2H₂O
MDL Number:MFCD00014234
:484.93 g/mol (448.90 anhy)
Boiling Pt:308 °C (1013 hPa)
Melting Pt:389…393 °C
Density:3,59 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-152-5
:GHS07
Related Information
Frequently Bought Together