Order Entry
Czech Republic
ContactUsLinkComponent
Lithium tri-tert-butoxyaluminum hydride ≥94%
Flammable
Corrosive
Lithium tri-tert-butoxyaluminum hydride ≥94%
Catalog # 30716.09
CAS Number:  
Flammable
Corrosive
Lithium tri-tert-butoxyaluminum hydride ≥94%
Catalog # 30716.09
CAS Number:  

Specifications

  • Pk:
    10 g

Specifications

About this item

 :
 :254.27 g/mol
Melting Pt:∼ 319 °C (dec.)
Density:1.03
Storage Temperature:Ambient
EINECS:241-490-8
ADR:4.3,I
 :GHS02
 :GHS05
CAS Number:17476-04-9
Formula:C₁₂H₂₈AlLiO₃
MDL Number:MFCD00011532