
n-Butyl phosphate mixture of mono-n-butyl and di-n-butyl
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52933-01-4
:
Formula:[CH₃(CH₂)₃O]₂P(O)OH/[CH₃(CH₂)₃O]P(O)(OH)₂
MDL Number:MFCD00013947
Boiling Pt:>124 °C (dec.)
Melting Pt:–60 °C
Density:1.123
Flash Pt:110 °C (230 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS05
Related Information
Frequently Bought Together