
Lithium tetrachloropalladate(II)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15525-45-8
:
Formula:Cl₄Li₂Pd
MDL Number:MFCD00011082
:262.1 g/mol
EINECS:239-567-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together