cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate (≥19% Ru)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15746-57-3
:
Formula:C₂₀H₂₀Cl₂N₄O₂Ru
MDL Number:MFCD00015504
:520.376 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together