
Potassium tetrabromoaurate(III) dihydrate (≥33% Au) ≥99.99% (metals basis), Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14323-32-1
:
Formula:KAuBr₄.2H₂O
MDL Number:MFCD00049650
:591.74 g/mol
EINECS:238-268-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together