

Ruthenium(III) nitrosylnitrate solution (1.5% Ru) (w/v)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34513-98-9
:
Formula:N₄O₁₀Ru
MDL Number:MFCD00016313
:317.09 g/mol
Density:1.07
Storage Temperature:Ambient
EINECS:252-068-8
UN:3093
ADR:8,II
:GHS05
:GHS03
Related Information
Frequently Bought Together