
Tetraammineplatinum(II) chloride monohydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13933-33-0
:
Formula:Cl₂H₁₄N₄OPt
MDL Number:MFCD00149947
:352.123 g/mol
Melting Pt:250 °C (dec.)
Density:2.74
EINECS:237-706-5
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together