
Diethyl (trichloromethyl)phosphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:866-23-9
:
Formula:C₅H₁₀Cl₃O₃P
MDL Number:MFCD00013666
:255.46 g/mol
Boiling Pt:128 °C (14 mmHg)
Density:1.3670
Storage Temperature:Ambient
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together