Anti-MUC5B Goat Polyclonal Antibody
Catalog # EVBIEB11765
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:mucin 5, subtype B, tracheobronchial
- Antigen symbol:MUC5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Format:affinity purified
- Form:liquid
- Antigen synonyms:AV085033|mucin 5b|MUC5|A130042M24|MUC9|2300002I04Rik
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:CQPQCQWTKWIDVDY
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Specifications
About this item
Western Blot: Not yet tested - our routinely used western blotting protocol does not allow detection of proteins as large as the calculated size of 515kDa according to NP_083077.2. Therefore we cannot recommend an optimal concentration and the antibody i
Type: Primary
Antigen: MUC5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat