Anti-PML Mouse Monoclonal Antibody [clone: PG-M3]
Catalog # SANTSC-966
Supplier: Santa Cruz Biotechnology
Specifications
- Antibody type:Primary
- Antigen name:promyelocytic leukemia
- Antigen symbol:PML
- Clonality:Monoclonal
- Clone:PG-M3
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Gene ID:5371
- Antigen synonyms:TRIM19|RNF71|PP8675|MYL
- Molecular weight:78/97 kDa
- Storage temperature:+4 °C
- Size:200 µg/ml
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: PML (promyelocytic leukemia)
Clonality: Monoclonal
Clone: PG-M3
Conjugation:
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human