Order Entry
Czech Republic
ContactUsLinkComponent
Anti-S100A11 Rabbit Polyclonal Antibody
Anti-S100A11 Rabbit Polyclonal Antibody
  BSBTPB9818
undefined
Anti-S100A11 Rabbit Polyclonal Antibody
  BSBTPB9818

 

  • Antibody type:
    Primary
  • Antigen name:
    S100 Calcium Binding Protein A11
  • Antigen symbol:
    S100A11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6282
  • Antigen synonyms:
    MLN70|HEL-S-43|S100C
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: S100A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human