ContactUsLinkComponent
BOD bottles, disposable
  HACH2943100
 :  Hach
undefined
BOD bottles, disposable
  HACH2943100
 :  Hach
  • Capacity:
    300 ml
  • Description:
    BOD bottle
  • Pk:
    100
PET, clear.
  • No cleaning required
  • Unbreakable and recyclable
  • Oxygen will not diffuse in or out of sample
These disposable bottles are fresh and ready to use.
 : EPA compliant.
 : Stoppers are sold separately.