Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KIT Mouse Monoclonal Antibody [clone: 566CT8.5.4]
Anti-KIT Mouse Monoclonal Antibody [clone: 566CT8.5.4]
Catalog # ABGEAM2137B
Supplier:  Abgent
Anti-KIT Mouse Monoclonal Antibody [clone: 566CT8.5.4]
Catalog # ABGEAM2137B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
  • Antigen symbol:
    KIT
  • Clonality:
    Monoclonal
  • Clone:
    566CT8.5.4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3815
  • Antigen synonyms:
    SCFR|CD117|PBT|C-Kit
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    109.865 kDa
  • Sequence:
    Recombinant Protein
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    9.500
  • Shipping temperature:
    4 °C
  • Immunogen:
    This KIT Antibody is generated from mouses immunized with human KIT recombinant protein.
  • Purification:
    Euglobin precipitation
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: 566CT8.5.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human