Dropping bottles, glass, with interchanging glass stopper
VTRB632426001500
: Kavalier
- Type:NS 29/32
- Capacity:500 ml
- Seq:03
- Pk:1
Soda-Lime glass, clear, SIMAX®.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
With interchanging glass hollow stopper.