ContactUsLinkComponent
UHPLC columns, Hypersil GOLD™ Vanquish™ C18
UHPLC columns, Hypersil GOLD™ Vanquish™ C18
Certificates
UHPLC columns, Hypersil GOLD™ Vanquish™ C18
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 554-5322
  • 554-5321
  • 554-5324
  • 554-5323
  • Carbon load:
  • 11%
  • 11%
  • 11%
  • 11%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • pH range:
  • 1 - 11
  • 1 - 11
  • 1 - 11
  • 1 - 11
  • Surface area:
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • 220 m²/g
  • Temperature limit:
  • 60 °C
  • 60 °C
  • 60 °C
  • 60 °C
  • USP code:
  • L1
  • L1
  • L1
  • L1