ContactUsLinkComponent
Quartz microfibre filters, Munktell
undefined
Quartz microfibre filters, Munktell
  • Catalog No:
  • BINZ420205
  • 537-0039
  • 537-0041
  • 537-0037
  • 537-0038
  • Grade:
  • MK 5
  • MK 5
  • MK 5
  • MK 5
  • MK 5
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,42 mm
  • 0,42 mm
  • 0,42 mm
  • 0,42 mm
  • 0,42 mm
  • Max. temperature:
  • 500 °C
  • 500 °C
  • 500 °C
  • 500 °C
  • 500 °C
Frequently Bought Together