ContactUsLinkComponent
Solution, cobalt chloride, for spectrophotometer
Solution, cobalt chloride, for spectrophotometer
Catalog # HACH1422249
Supplier:  Hach
CAS Number:  
undefined
Solution, cobalt chloride, for spectrophotometer
Catalog # HACH1422249
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    Cobalt chloride solution, 500 ml
Specifications
About this item
Cobalt chloride solution for spectrophotometer linearisation.