Order Entry
Czech Republic
ContactUsLinkComponent
Alkalinity standard solution, 25000 mg/L CaCO₃ (NIST)
Alkalinity standard solution, 25000 mg/L CaCO₃ (NIST)
Catalog # 705-2018
Supplier:  Hach
CAS Number:  
undefined
Alkalinity standard solution, 25000 mg/L CaCO₃ (NIST)
Catalog # 705-2018
Supplier:  Hach
CAS Number:  

Specifications

  • Concentration:
    25000 mg/l as CaCO3
  • Pack type:
    10 ml Voluette ampoules
  • Pk:
    16
  • Description:
    Alkalinity standard solution

Specifications

About this item

Standard solution for accuracy checks in alkalinity determinations.

  • Simple accuracy checks
  • Minimise waste
  • Reliable results

Voluette and PourRite ampule standards are designed to work directly with standard additions accuracy checks in Hach procedures and photometer software. Use the Tensette pipet to add precise spikes of standard and check percent recovery, to confirm analytical system performance.