Order Entry
Czech Republic
ContactUsLinkComponent
Carbon dioxide standard solution
Carbon dioxide standard solution
Catalog # HACH226142
Supplier:  Hach
CAS Number:  
undefined
Carbon dioxide standard solution
Catalog # HACH226142
Supplier:  Hach
CAS Number:  

Specifications

  • Concentration:
    100 mg/l as CO2
  • Pk:
    100 mL
  • Description:
    Carbon dioxide standard solution, voluette ampoule, 100 ml
  • Seq:
    01

Specifications

About this item

Standard solution for accuracy checks in carbon dioxide determinations (Potassium hydrogen phthalate solution).