ContactUsLinkComponent
Nickel standard solution
Nickel standard solution
  HACH1417642
 :  Hach
 :  
undefined
Nickel standard solution
  HACH1417642
 :  Hach
 :  
  • Concentration:
    1000 mg/l as Ni
  • Pk:
    100 mL
  • Description:
    Nickel standard solution (NIST)
Standard solution for accuracy checks in nickel determinations.