Order Entry
Czech Republic
ContactUsLinkComponent
Spectromelt® C 10 grinding and tabletting aid, for XRF (X-ray fluorescence), Supelco®
Spectromelt® C 10 grinding and tabletting aid, for XRF (X-ray fluorescence), Supelco®
Catalog # 1.13990.5000
Supplier:  Merck
CAS Number:  
undefined
Spectromelt® C 10 grinding and tabletting aid, for XRF (X-ray fluorescence), Supelco®
Catalog # 1.13990.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • IR-spectrum:
    passes test
  • Al (Aluminium):
    ≤ 0,001
  • Ba (Barium):
    ≤ 0,001
  • Ca (Calcium):
    ≤ 0,001
  • Co (Cobalt):
    ≤ 0,001
  • Cr (Chromium):
    ≤ 0,001
  • Cu (Copper):
    ≤ 0,001
  • Fe (Iron):
    ≤ 0,001
  • K (Potassium):
    ≤ 0,001
  • Mn (Manganese):
    ≤ 0,001
  • Mo (Molybdenum):
    ≤ 0,001
  • Ni (Nickel):
    ≤ 0,001
  • Pb (Lead):
    ≤ 0,001
  • Si (Silicon):
    ≤ 0,001
  • Sr (Strontium):
    ≤ 0,001
  • Ti (Titanium):
    ≤ 0,001
  • V (Vanadium):
    ≤ 0,001

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100