Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Balances and Scales
Accessories for scales, food portioning, FD series
Accessories for scales, food portioning, FD series
Accessories for scales, food portioning, FD series
Catalog # 611-2413
Supplier:
OHAUS
Expand image
Accessories for scales, food portioning, FD series
Catalog # 611-2413
Supplier:
OHAUS
Specifications
Description:
Density determination kit for Explorer®/Adventurer® balances (0,01, 0,1 and 1 mg models only)
Seq:
59
Pk:
1
Specifications
About this item
Product Details
Product Details