Starch, soluble (from potato), powder, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-686-4
Specification Test Results
- Meets ACS Reagent Chemical Requirements:5.0 - 7.0
- For Laboratory,Research,or Manufacturing Use:≤0.4%
- Solubility:Passes Test
- pH of 2% Solution at 25°C:5.0 - 7.0
- Residue after Ignition:≤0.4 %
- Sensitivity:Passes Test
Specification Test Results
Frequently Bought Together