

Potassium iodate 99.4-100.4%, BAKER ANALYZED® ACS, J.T.Baker®
3156-04
: Avantor
:
- Pack type:Poly bottle, white, blue T/E cap
- Pk:125 g
- Seq:3445
- Assay (KIO₃):99.4 - 100.4 %
- Insoluble Matter:≤0.005 %
- pH of 5% Solution at 25°C:5.0 - 8.0
- Chloride and Bromide (as Cl):≤0.01 %
- Iodide (I):≤0.001 %
- Nitrogen Compounds (as N):≤0.005 %
- Sulfate (SO₄):≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Sodium (Na):≤0.005 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
:
:214 g/mol
:13,07726
:560 °C
:3.98 g/cm³ (20 °C)
:Ambient
:231-831-9
:1479
:5.1,II
:GHS03
:GHS07
:7758-05-6
:KlO₃
:MFCD00011406