Order Entry
Czech Republic
ContactUsLinkComponent
Potassium bromide ≥99%, BAKER INSTRA-ANALYZED® for spectroscopy, J.T.Baker®
Potassium bromide ≥99%, BAKER INSTRA-ANALYZED® for spectroscopy, J.T.Baker®
Catalog # 2961-05
Supplier:  Avantor
CAS Number:  
undefined
Potassium bromide ≥99%, BAKER INSTRA-ANALYZED® for spectroscopy, J.T.Baker®
Catalog # 2961-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (KBr) (by Ag titrn):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • Loss on Drying at 105°C:
    ≤0.05 %
  • pH of 5% Solution at 25°C:
    5.0 - 7.5
  • Nitrogen Compounds (as N):
    ≤0.0005 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Suitability for Infrared Analysis:
    Passes Test

Specification Test Results

About this item

 :
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
 :119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
 :GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358