Order Entry
Czech Republic
ContactUsLinkComponent
4-(Dimethylamino)benzaldehyde, BAKER ANALYZED® ACS, J.T.Baker®
4-(Dimethylamino)benzaldehyde, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # J418-05
Supplier:  Avantor
CAS Number:  
undefined
4-(Dimethylamino)benzaldehyde, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # J418-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    73 - 75 °C
  • For Laboratory,Research,or Manufacturing Use:
    73 - 75 °C
  • Melting Point:
    73 - 75 °C
  • Solubility in HCl:
    Passes Test
  • Solubility in Alcohol:
    Passes Test
  • Color of HCl Solution:
    Passes Test
  • Color of Alcohol Solution(APHA):
    ≤60
  • Residue after Ignition:
    ≤0.10 %

Specification Test Results

About this item

 :
 :149.19 g/mol
Merck Index:13,03257
Boiling Pt:176…177 °C (1013 hPa)
Melting Pt:72 °C
Density:1.0254 g/cm³ (99.9 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:202-819-0
CAS Number:100-10-7
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381