Order Entry
Czech Republic
ContactUsLinkComponent
Oxalic acid dihydrate 99.5-102.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Oxalic acid dihydrate 99.5-102.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0230-07
Supplier:  Avantor
CAS Number:  
undefined
Oxalic acid dihydrate 99.5-102.5%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0230-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    12 kg
  • Seq:
    4187
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.5 - 102.5%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (HOCOCOOH2H₂O) (by KMnO₄ titrn):
    99.5 - 102.5 %
  • Insoluble Matter:
    ≤ 0.005 %
  • Residue after Ignition:
    ≤ 0.010 %
  • Chloride (Cl):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 0.002 %
  • Calcium (Ca)(by FES):
    ≤ 0.001 %
  • Nitrogen Compounds (as N):
    ≤ 0.001 %
  • Substances Darkened by Hot H₂SO₄:
    Passes Test
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 2.0 ppm

Specification Test Results

About this item

Synonyms: Ethanedioic acid dihydrate
 :
 :126.07 g/mol
Melting Pt:101
EINECS:205-634-3
UN:3261
REACH:01-2119534576-33
 :GHS07
Storage Temperature:Ambient
CAS Number:6153-56-6
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102
Boiling Pt:149…160 °C (1013 hPa)
Density:1.65 g/cm³ (20 °C)
ADR:8 III
Merck Index:13,06980