Order Entry
Czech Republic
ContactUsLinkComponent
Amyl acetate mixture of isomers, purified, J.T.Baker®
Flammable
Amyl acetate mixture of isomers, purified, J.T.Baker®
Catalog # 9094-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Amyl acetate mixture of isomers, purified, J.T.Baker®
Catalog # 9094-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Identification (by IR):
    Passes Test

Specification Test Results

About this item

 :
 :130.19 g/mol
Boiling Pt:149 °C (1013 hPa)
Melting Pt:–71 °C
Density:0.876 g/cm³ (20 °C)
Flash Pt:23 °C (closed cup)
Storage Temperature:Ambient
EINECS:211-047-3
UN:1104
ADR:3,III
 :GHS02
CAS Number:628-63-7
Formula:C₇H₁₄O₂
MDL Number:MFCD00009500