Extraction systems and accessories for industrial welding
Supplier: asecos
Certificates
About this item
These extraction systems are ideal for use in narrow places such as welding booths or at bank work. They have externally mounted brackets for natural, straight run of the exhaust air.
- Integrated throttle
- Extremely light and stable position
- Gas spring for balancing the dead weight
- Low pressure drop for effective extraction
Frequently Bought Together