Order Entry
Czech Republic
ContactUsLinkComponent
proNGF Rapid™ ELISA Kit: Rat and Mouse
proNGF Rapid™ ELISA Kit: Rat and Mouse
Supplier:  Biosensis
Certificates
undefined
proNGF Rapid™ ELISA Kit: Rat and Mouse
Supplier:  Biosensis

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Rat
    Mouse
  • Target protein:
    proNGF
  • Cross reactivity:
    Reacts with human proNGF (about 20% reactivity). Does not cross-react with proBDNF and mature NGF. Mature NGF spiked into brain homogenate does not interfere with proNGF quantification.
  • Detection range:
    0,156 – 10 ng/ml
  • Sensitivity:
    Typical limit of detection (LOD) for mouse proNGF is less than 50 pg/ml determined as blank value plus 3x standard deviation of blank OD (n=10).

Specifications

Frequently Bought Together