- Pk:192 Tests
- Conjugate ELISA:HRP
- Format:Pre-coated
- Host:
- Primary antibody reactivity:Human
- Target protein:Neurotrophins: BDNF, NGF, NT3, NT4/5
- Description:Multi-Neurotrophin Rapid Screening ELISA Kit: Human, 2×96-well plate
- Environmentally Preferable:
- Sample type:Cell culture supernatants, lysates and brain extracts
- Cross reactivity:The antibodies used in this kit detect the mouse and rat form of each neurotrophin, but with varying efficiency depending on the target protein.
- Detection method:Photometric (450 nm)
- Specificity:Each kit has been tested for cross-reactivity with other closely related neurotrophins and no cross-reactivity has been observed at 25 ng/ml of each neurotrophin tested.
- Detection range:NGF (3,9 - 250 pg/ml); BDNF (7,8 - 500 pg/ml); NT3 (15,6 - 1000 pg/ml); NT4/5 (15,6 - 1000 pg/ml)
- Sensitivity:Typical LODs achieved based on 150% of background OD: NGF: 3 pg/ml BDNF: 1 pg/ml NT3: 4 pg/ml NT4/5: 10 pg/ml
The Biosensis Multi-Neurotrophin Rapid™ Screening ELISA Kit has been designed to allow rapid screening and quantification of NGF, BDNF, NT3 and NT4/5 in cell culture supernatants, lysates, serum, plasma (EDTA and citrate) and brain extracts only if used as directed.
This two-plate kit consists of four sets of 6 strips for each Neurotrophin, allowing for the assay of 16 samples per Neurotrophin tested and a full range of standards, all run in duplicate. It provides the identical sensitivities and ranges that are achieved in the complete, individual ELISA kits, thus allowing easy progression into the larger individual assay sizes when needed.
The Multi-Neurotrophin Rapid™ Screening ELISA Kit presents a cost effective way to quickly screen multiple samples, which can then be published or used prior to a more extensive analysis with the individual kits. The kit comes complete with all detection reagents and is ready to use. Individual coated strips are provided and each set of standards and detection antibodies are colour-coded.
Please refer to the kit protocol for specific use instructions for each substrate application, in particular human serum and plasma samples.