ContactUsLinkComponent
Affinity chromatography columns, tagged package, GST
Affinity chromatography columns, tagged package, GST
Catalog # 29-0588-04
Supplier:  Cytiva
undefined
Affinity chromatography columns, tagged package, GST
Catalog # 29-0588-04
Supplier:  Cytiva
Specifications
  • Description:
    Tagged package, GST
  • Pk:
    1 KIT
Specifications
About this item
The GST package is designed for easy purification of GST-tagged proteins.
The GSTrap™ 4B column (1×1 ml) has a high binding capacity for GST-tagged proteins, giving you higher yields. A HiTrap™ desalting column (1×5 ml) is provided for rapid buffer exchange.
Delivery: The kit contains one of each GSTrap™ 4B (1×1 ml) and HiTrap™ Desalting (1×5 ml) columns.