Order Entry
Czech Republic
ContactUsLinkComponent
Potassium hydroxide 45.0 - 46.5% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Corrosive
Potassium hydroxide 45.0 - 46.5% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Potassium hydroxide 45.0 - 46.5% in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    45.0 - 46.5 %
  • Assay (KOH):
    45.0 - 46.5 %
  • Potassium Carbonate (K₂CO₃):
    ≤0.2 %
  • Chloride (Cl):
    ≤0.003 %
  • Sulfate (SO₄):
    ≤0.001 %
  • Ammonium Hydroxide Precipitate:
    ≤0.005 %
  • Sodium (Na):
    ≤0.02 %
  • Nitrogen Compounds (as N):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤2 ppm
  • Heavy Metals (as Ag):
    ≤5 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.050 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm
  • Trace Impurities - Nickel (Ni):
    ≤5.000 ppm

Specification Test Results

Related Information

Frequently Bought Together