Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Katalogové číslo 786-R40
Dodavatel:  G-Biosciences
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Katalogové číslo 786-R40
Dodavatel:  G-Biosciences

Specifikace

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Rat
  • Seq:
    03
  • Size:
    2 ml
  • Pk:
    2 mL

Specifikace

O tomto produktu