Anti-IgG Rabbit Antibody (HRP (Horseradish Peroxidase))
Catalog # 786-R42
Supplier: G-Biosciences
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Goat
- Seq:05
- Pk:1,5 mL