Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
786-R38
- Antibody type:Secondary
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Mouse
- Seq:01
- Pk:2 mL