Anti-PDK4 Rabbit Polyclonal Antibody
BSBTPB9773
- Antibody type:Primary
- Antigen symbol:PDK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5166
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 4, mitochondrial(PDK4) detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: PDK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat