ContactUsLinkComponent
Safety drum vents
Safety drum vents
Catalog # 119-0234
undefined
Safety drum vents
Catalog # 119-0234
Specifications
  • Description:
    Brass vertical vent for petroleum based applications, with manual vacuum relief. Type: Manual vacuum relief, 2" NPS/NPT
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
These safety drum vents provide automatic pressure relief for drums between 0,20 bar but less than 0,55 bar. Depending on the model, vacuum relief is either automatic (on horizontally installed vents) or manual (on vertically installed vents).
  • Vents can prevent escape of flammable vapour in closed position
  • Includes flame arrester for added safety