Silicic acid hydrate, extra pure, precipitated light, Supelco®
Catalog # 1.00657.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Fibre carton
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:1710 °C
- Molar Mass:60.08 g/mol
- Bulk Density:60 kg/m3
- Density:2 g/cm3 (20 °C)
- Synonyms:DAB
- Hill Formula:O2Si *x H2O
- Chemical Formula:SiO2 * x H2O
- Storage temperature:+5...+30 °C
- Identity:Passes test
- Assay:99,0 - 100,5%
- Chloride (Cl):≤0,05%
- Heavy metals (as Pb):≤0,0025%
- Fe (Iron):≤0,03%
- Particle size:0,1 mm
- Loss on ignition:≤8,5% at 900 °C
- Sulphate (SO4):≤0,5%
- pH value:3,0 - 8,0
Specification Test Results
About this item
:
:60.09 g/mol (anhy)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:10279-57-9
:63231-67-4
Formula:H₂O₃Si
MDL Number:MFCD00054122