Order Entry
Czech Republic
ContactUsLinkComponent
Clean-Lab, Q Path® for paraffin removal
Flammable
Health
Clean-Lab, Q Path® for paraffin removal
Catalog # 10047400.
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Clean-Lab, Q Path® for paraffin removal
Catalog # 10047400.
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Aromatics (UV) (max. 100 ppm):
    Passes test
  • Boiling point (begin) (min.173°C):
    Passes test
  • Bromine (per 100 g) (max. 50 ppm):
    Passes test
  • Colouration (Saybolt) (D6045) (min. 30):
    Passes test
  • Density (15°C) (0.752-0.772 kg/l):
    Passes test
  • Distillation range (50 %) (177-187°C):
    Passes test
  • Dry point (boiling range) (max.193°C):
    Passes test
  • Flash point (56-63°C):
    Passes test
  • S (Sulphur) (max. 1 ppm):
    Passes test

Specification Test Results

About this item

 :
Flash Pt:49 °C (closed cup)
UN:3295
ADR:3,III
 :GHS02
 :GHS08
 :1
CAS Number:90622-57-4