

Triallyl isocyanurate for synthesis, Sigma-Aldrich®
Catalog # 8.16113.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:3.7 g/l (30 °C)
- Melting Point:25 °C
- Molar Mass:249.27 g/mol
- Boiling Point:150 °C (5.3 hPa)
- Vapor Pressure:3.5 hPa (143 °C)
- Flash Point:160 °C
- Density:1.16 g/cm3 (20 °C)
- Viscosity Kinematic:78.2 mm2/s (30 °C)
- Storage:+15...+25 °C
- Assay:≥98,0% (a/a)
- Identity:Passes test
Specification Test Results
About this item
:
:249.27 g/mol
:M-Clarity™ quality level = MQ200
EINECS:213-834-7
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:1025-15-6
Formula:C₁₂H₁₅N₃O₃
MDL Number:MFCD00006554