Anti-COPS7A Rabbit Polyclonal Antibody
Catalog # BOSSBS-9124R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:COP9 signalosome subunit 7A
- Antigen symbol:COPS7A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:50813
- Antigen synonyms:Signalosome subunit 7a|DERP10|COP9 signalosome complex subunit 7a|JAB1 containing signalosome subunit 7a|SGN7a|CSN7A_HUMAN.|Dermal papilla derived protein 10|CSN7A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
COPS7A is a component of the COP9 signalosome complex (CSN), a complex involved in various cellular and developmental processes. The CSN complex is an essential regulator of the ubiquitin (Ubl) conjugation pathway by mediating the deneddylation of the cullin subunits of SCF-type E3 ligase complexes, leading to decrease the Ubl ligase activity of SCF-type complexes such as SCF, CSA or DDB2.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: COPS7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: