Anti-SMS Rabbit Polyclonal Antibody
Katalogové číslo BOSSBS-8681R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Spermine Synthase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Antigen synonyms:MRSR|SMS|SPSY_HUMAN|Snyder Robinson X linked mental retardation syndrome|SRS.|Spermine synthase|Spermidine aminopropyltransferase|SPMSY|SpS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Spermine synthase catalyzes the production of spermine from spermidine. Spermine, a polyamine ubiquitously present in most organisms, is essential for normal cell growth and differentiation. Because absence of spermine increases sensitivity of cells to anti-tumor agents, spermine synthase (and other polyamine biosynthesis) is an attractive target for anti-neoplastic therapy.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: SMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat