Specifications
- Antibody type:Primary
- Antigen name:Arrestin domain containing 3
- Antigen symbol:ARRDC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:57561
- Antigen synonyms:TLIMP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
The arrestins are a family of proteins that are important for regulating signal transduction within cells. Arrestins are part of a conserved two step mechanism for regulating the activity of G-protein coupled receptors (GPCRs). In response to a stimulus, GPCRs activate a heterotrimeric G protein. In order to turn off this response, or adapt to a constant stimulus, activated receptors need to be silenced. The first step is phosphorylation by a class of serine/threonine kinases called G protein coupled receptor kinases (GRKs). This phosphorylation specifically marks the activated receptor for arrestin binding. Once arrestin is bound to the receptor it is unable to signal further. Recent research continues to expand the known actions of arrestins, which can bind to other classes of receptors and can directly activate signaling pathways on their own. Different arrestins (visual arrestin (or Arrestin 1), beta-arrestin 1 (or Arrestin 2) and beta-arrestin 2 (or Arrestin 3) can reduce the activity of their target GPCRs in several different ways.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ARRDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat